Anti-ACTR3B Rabbit Recombinant Polyclonal Antibody
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ACTR3B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ACTR3B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77840-082
- 77840-083
- Antigen Symbol:
- ACTR3B
- ACTR3B
- Antigen Name:
- Actin-related protein 3B protein (189-418AA)
- Actin-related protein 3B protein (189-418AA)
- Antigen Synonyms:
- ACTR3B|ARP11|ARP4Actin-related protein 3B|ARP3-beta|Actin-like protein 3B|Actin-related protein ARP4
- ACTR3B|ARP11|ARP4Actin-related protein 3B|ARP3-beta|Actin-like protein 3B|Actin-related protein ARP4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P1U1
- Q9P1U1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Product Family Options
- SizePackaging
- : A69163-100UL
Specifications
Cat. No.77840-082Antigen SymbolACTR3BAntigen NameActin-related protein 3B protein (189-418AA)Antigen SynonymsACTR3B|ARP11|ARP4Actin-related protein 3B|ARP3-beta|Actin-like protein 3B|Actin-related protein ARP4ClonalityRecombinant PolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene IDQ9P1U1IsotypeIgGELISAYesImmunoChemistryYesFormLiquidPurificationAntigen affinity purifiedStorage BufferPBS, 0.02% sodium azide, 50% glycerol, pH 7.3.Storage TemperatureUpon receipt, store at –20 or –80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice - : A69163-50UL
Specifications
Cat. No.77840-083Antigen SymbolACTR3BAntigen NameActin-related protein 3B protein (189-418AA)Antigen SynonymsACTR3B|ARP11|ARP4Actin-related protein 3B|ARP3-beta|Actin-like protein 3B|Actin-related protein ARP4ClonalityRecombinant PolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene IDQ9P1U1IsotypeIgGELISAYesImmunoChemistryYesFormLiquidPurificationAntigen affinity purifiedStorage BufferPBS, 0.02% sodium azide, 50% glycerol, pH 7.3.Storage TemperatureUpon receipt, store at –20 or –80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice



