Anti-THOC6 Rabbit Recombinant Polyclonal Antibody
Catalog # 77831-992
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:THO complex subunit 6 homolog protein (56-315AA)
- Antigen Symbol:THOC6
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q86W42
- Antigen Synonyms:WDR58|PSEC0006|THO complex subunit 6 homolog|Functional spliceosome-associated protein 35|fSAP35|WD repeat-containing protein 58
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: THOC6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human