Anti-GNPDA2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNPDA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-585
- 77833-584
- Antigen Symbol:
- GNPDA2
- GNPDA2
- Antigen Name:
- Glucosamine-6-phosphate isomerase 2 protein (28-154AA)
- Glucosamine-6-phosphate isomerase 2 protein (28-154AA)
- Antigen Synonyms:
- GlcN6P deaminase 2| Glucosamine 6 phosphate deaminase 2| Glucosamine 6 phosphate isomerase 2| Glucosamine 6 phosphate isomerase SB52| Glucosamine-6-phosphate deaminase 2| Glucosamine-6-phosphate isomerase 2| Glucosamine-6-phosphate isomerase SB52| GNP2| GNPDA 2| Gnpda2| GNPI2_HUMAN| SB52
- GlcN6P deaminase 2| Glucosamine 6 phosphate deaminase 2| Glucosamine 6 phosphate isomerase 2| Glucosamine 6 phosphate isomerase SB52| Glucosamine-6-phosphate deaminase 2| Glucosamine-6-phosphate isomerase 2| Glucosamine-6-phosphate isomerase SB52| GNP2| GNPDA 2| Gnpda2| GNPI2_HUMAN| SB52
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDQ7
- Q8TDQ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together