Anti-RIT2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RIT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-102
- 77835-101
- Antigen Symbol:
- RIT2
- RIT2
- Antigen Name:
- GTP-binding protein Rit2 protein (1-150AA)
- GTP-binding protein Rit2 protein (1-150AA)
- Antigen Synonyms:
- GTP binding protein Rit2|GTP binding protein Roc2|GTP-binding protein Rit2|Ras like protein expressed in neurons|Ras like without CAAX 2|Ras like without CAAX protein 2|Ras-like protein expressed in neurons|Ras-like without CAAX protein 2|RIBA|Ric (Drosophila) like|Ric like|Ric like, expressed in neurons|RIN|RIT2|RIT2_HUMAN|ROC2
- GTP binding protein Rit2|GTP binding protein Roc2|GTP-binding protein Rit2|Ras like protein expressed in neurons|Ras like without CAAX 2|Ras like without CAAX protein 2|Ras-like protein expressed in neurons|Ras-like without CAAX protein 2|RIBA|Ric (Drosophila) like|Ric like|Ric like, expressed in neurons|RIN|RIT2|RIT2_HUMAN|ROC2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99578
- Q99578
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together