Anti-SGCG Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SGCG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-418
- 77823-417
- Antigen Symbol:
- 35 kDa dystrophin associated glycoprotein;35 kDa dystrophin-associated glycoprotein;35DAG;35kD dystrophin associated glycoprotein;35kDa dystrophin-associated glycoprotein;A4;DAGA4;DMDA;DMDA1;Gamma SG;Gamma-sarcoglycan;Gamma-SG;LGMD2C;MAM;MGC130048;Sarcoglycan gamma;SCARMD2;SCG3;SGCG;SGCG_HUMAN;TYPE
- 35 kDa dystrophin associated glycoprotein;35 kDa dystrophin-associated glycoprotein;35DAG;35kD dystrophin associated glycoprotein;35kDa dystrophin-associated glycoprotein;A4;DAGA4;DMDA;DMDA1;Gamma SG;Gamma-sarcoglycan;Gamma-SG;LGMD2C;MAM;MGC130048;Sarcoglycan gamma;SCARMD2;SCG3;SGCG;SGCG_HUMAN;TYPE
- Antigen Name:
- Gamma-sarcoglycan protein (108-221AA)
- Gamma-sarcoglycan protein (108-221AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13326
- Q13326
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together