Anti-C1QTNF5 Guinea pig Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: C1QTNF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77806-563
- 77806-562
- Antigen Symbol:
- C1QTNF5
- C1QTNF5
- Antigen Name:
- Synthetic peptide of C1QTNF5
- Synthetic peptide of C1QTNF5
- Antigen Synonyms:
- CTRP5|UNQ303/PRO344|Complement C1q tumor necrosis factor-related protein 5
- CTRP5|UNQ303/PRO344|Complement C1q tumor necrosis factor-related protein 5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Guinea pig
- Guinea pig
- Gene ID:
- Q9BXJ0
- Q9BXJ0
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together