Anti-VAC14 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: VAC14
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-521
- 77821-520
- Antigen Symbol:
- VAC14
- VAC14
- Antigen Name:
- Protein VAC14 homolog protein (333-523AA)
- Protein VAC14 homolog protein (333-523AA)
- Antigen Synonyms:
- ArPIKfyve|FLJ10305|FLJ36622|FLJ46582|MGC149815|MGC149816|OTTHUMP00000174908|Protein VAC14 homolog|Tax1 (human T cell leukemia virus type I) binding protein 1|Tax1 (human T cell leukemia virus type I) binding protein 2|Tax1 binding protein 2|Tax1-binding protein 2|TAX1BP2|TRX|VAC 14|vac14|Vac14 homolog (S. cerevisiae)|Vac14 homolog|VAC14_HUMAN
- ArPIKfyve|FLJ10305|FLJ36622|FLJ46582|MGC149815|MGC149816|OTTHUMP00000174908|Protein VAC14 homolog|Tax1 (human T cell leukemia virus type I) binding protein 1|Tax1 (human T cell leukemia virus type I) binding protein 2|Tax1 binding protein 2|Tax1-binding protein 2|TAX1BP2|TRX|VAC 14|vac14|Vac14 homolog (S. cerevisiae)|Vac14 homolog|VAC14_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q08AM6
- Q08AM6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together