Anti-TFPT Rabbit Polyclonal Antibody
Catalog # 77802-351
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:TFPT_HUMAN
- Antigen Symbol:TFPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P0C1Z6
- Antigen Synonyms:Amida|INO80 complex subunit F|INO80F|Protein FB1|TCF3 (E2A) fusion partner (in childhood Leukemia)|TCF3 fusion partner|TFPT|TFPT_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TFPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat