Anti-U2AF2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: U2AF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77803-797
- 77811-797
- Antigen Symbol:
- U2AF2
- U2AF2
- Antigen Name:
- Splicing factor U2AF 65 kDa subunit protein (124-246AA)
- Splicing factor U2AF 65 kDa subunit protein (124-246AA)
- Antigen Synonyms:
- hU2AF(65)|hU2AF65|Splicing factor U2AF 65 kDa subunit|U2 (RNU2) small nuclear RNA auxiliary factor 2|U2 auxiliary factor 65 kDa subunit|U2 small nuclear ribonucleoprotein auxiliary factor (65kD)|U2 small nuclear RNA auxiliary factor 2|U2 snRNP auxiliary factor large subunit|U2af2|U2AF2_HUMAN|U2AF65
- hU2AF(65)|hU2AF65|Splicing factor U2AF 65 kDa subunit|U2 (RNU2) small nuclear RNA auxiliary factor 2|U2 auxiliary factor 65 kDa subunit|U2 small nuclear ribonucleoprotein auxiliary factor (65kD)|U2 small nuclear RNA auxiliary factor 2|U2 snRNP auxiliary factor large subunit|U2af2|U2AF2_HUMAN|U2AF65
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P26368
- P26368
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together