Anti-USP30 Rabbit Recombinant Polyclonal Antibody
50 μg, Vial
Catalog # 77831-540
Supplier: AFG Bioscience
Supplier #: A59250-50UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type:
Antigen: USP30
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Specifications
- Size:50 μg
- Packaging:Vial
- Clonality:Recombinant Polyclonal
- Purification:>95%, Protein G purified
- Form:Liquid
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77831-540
- Reactivity:Mouse|Human
- Western Blot:Yes
- Shipping Temperature:Shipped on Blue ice
- Antigen Symbol:USP30
- Gene ID:Q70CQ3
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:Deubiquitinating enzyme 30|FLJ40511|Ub-specific protease 30|Ubiquitin carboxyl terminal hydrolase 30|Ubiquitin carboxyl-terminal hydrolase 30|Ubiquitin specific peptidase 30|Ubiquitin specific processing protease 30|Ubiquitin thioesterase 30|Ubiquitin thiolesterase 30|Ubiquitin-specific-processing protease 30|UBP30_HUMAN|usp30
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 30 protein (101-400AA)
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.



