Anti-DDR2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DDR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-216
- 77824-215
- Antigen Symbol:
- DDR2
- DDR2
- Antigen Name:
- Discoidin domain-containing receptor 2 protein (447-540AA)
- Discoidin domain-containing receptor 2 protein (447-540AA)
- Antigen Synonyms:
- CD167 antigen-like family member B|CD167b|CD167b antigen|Cell migration inducing protein 20|DDR 2||_HUMAN|Discoidin domain containing receptor 2|Discoidin domain receptor 2|Discoidin domain receptor family member 2|discoidin domain receptor tyrosine kinase 2|Discoidin domain-containing receptor 2|discoidin domain-containing receptor tyrosine kinase 2|Hydroxyaryl protein kinase|MIG20a|Migration inducing gene 16 protein|Neurotrophic tyrosine kinase|Neurotrophic tyrosine kinase receptor related 3|NTRKR 3|NTRKR3|Receptor protein tyrosine kinase TKT|Receptor protein-tyrosine kinase TKT|Receptor related 3|receptor-related 3|TKT|TYRO 10|TYRO10|Tyrosine kinase receptor related to neurotrophic TRK|Tyrosine protein kinase TYRO 10|Tyrosine protein kinase TYRO10|Tyrosine-protein kinase TYRO10|Tyrosylprotein kinase
- CD167 antigen-like family member B|CD167b|CD167b antigen|Cell migration inducing protein 20|DDR 2||_HUMAN|Discoidin domain containing receptor 2|Discoidin domain receptor 2|Discoidin domain receptor family member 2|discoidin domain receptor tyrosine kinase 2|Discoidin domain-containing receptor 2|discoidin domain-containing receptor tyrosine kinase 2|Hydroxyaryl protein kinase|MIG20a|Migration inducing gene 16 protein|Neurotrophic tyrosine kinase|Neurotrophic tyrosine kinase receptor related 3|NTRKR 3|NTRKR3|Receptor protein tyrosine kinase TKT|Receptor protein-tyrosine kinase TKT|Receptor related 3|receptor-related 3|TKT|TYRO 10|TYRO10|Tyrosine kinase receptor related to neurotrophic TRK|Tyrosine protein kinase TYRO 10|Tyrosine protein kinase TYRO10|Tyrosine-protein kinase TYRO10|Tyrosylprotein kinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16832
- Q16832
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together