Anti-L1TD1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: L1TD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-766
- 77828-765
- Antigen Symbol:
- L1TD1
- L1TD1
- Antigen Name:
- LINE-1 type transposase domain-containing protein 1 protein (665-865AA)
- LINE-1 type transposase domain-containing protein 1 protein (665-865AA)
- Antigen Synonyms:
- ECAT11|ES cell associated protein 11|ES cell-associated protein 11|FLJ10884|L1td1|LINE 1 type transposase domain containing 1|LINE 1 type transposase domain containing protein 1|LINE-1 type transposase domain-containing protein 1|LITD1_HUMAN|MGC133253|OTTHUMP00000010477|RP5-1155K23.3
- ECAT11|ES cell associated protein 11|ES cell-associated protein 11|FLJ10884|L1td1|LINE 1 type transposase domain containing 1|LINE 1 type transposase domain containing protein 1|LINE-1 type transposase domain-containing protein 1|LITD1_HUMAN|MGC133253|OTTHUMP00000010477|RP5-1155K23.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5T7N2
- Q5T7N2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together