Anti-TAF7L Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TAF7L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77827-040
- 77827-039
- Antigen Symbol:
- TAF7L
- TAF7L
- Antigen Name:
- Transcription initiation factor TFIID subunit 7-like protein (1-98AA)
- Transcription initiation factor TFIID subunit 7-like protein (1-98AA)
- Antigen Synonyms:
- TAF7L|TAF2Q|Transcription initiation factor TFIID subunit 7-like|Cancer/testis antigen 40|CT40|RNA polymerase II TBP-associated factor subunit Q|TATA box-binding protein-associated factor 50 kDa|Transcription initiation factor TFIID 50 kDa subunit
- TAF7L|TAF2Q|Transcription initiation factor TFIID subunit 7-like|Cancer/testis antigen 40|CT40|RNA polymerase II TBP-associated factor subunit Q|TATA box-binding protein-associated factor 50 kDa|Transcription initiation factor TFIID 50 kDa subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5H9L4
- Q5H9L4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together