Anti-UNC5B Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UNC5B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-016
- 77804-015
- Antigen Symbol:
- UNC5B
- UNC5B
- Antigen Name:
- Netrin receptor UNC5B protein (404-517AA)
- Netrin receptor UNC5B protein (404-517AA)
- Antigen Synonyms:
- 6330415E02Rik|A630020F16|D10Bwg0792e|Netrin receptor UNC5B|Netrin receptor UNC5B Precursor|p53 regulated receptor for death and life|p53 regulated receptor for death and life protein 1|p53-regulated receptor for death and life protein 1|p53RDL1|Protein unc-5 homolog 2|Protein unc-5 homolog B|Protein XUNC 5|Transmembrane receptor Unc5H2|Unc 5 homolog 2|Unc 5 homolog B|Unc 5 netrin receptor B|UNC 5B|Unc5 (C.elegans homolog) b|Unc5b|UNC5B_HUMAN|UNC5H2
- 6330415E02Rik|A630020F16|D10Bwg0792e|Netrin receptor UNC5B|Netrin receptor UNC5B Precursor|p53 regulated receptor for death and life|p53 regulated receptor for death and life protein 1|p53-regulated receptor for death and life protein 1|p53RDL1|Protein unc-5 homolog 2|Protein unc-5 homolog B|Protein XUNC 5|Transmembrane receptor Unc5H2|Unc 5 homolog 2|Unc 5 homolog B|Unc 5 netrin receptor B|UNC 5B|Unc5 (C.elegans homolog) b|Unc5b|UNC5B_HUMAN|UNC5H2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IZJ1
- Q8IZJ1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together