Anti-CHPF2 Rabbit Polyclonal Antibody
Catalog # 77806-251
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the N-terminal region of CSGlcA-T
- Antigen Symbol:CHPF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9P2E5
- Antigen Synonyms:Chondroitin glucuronyltransferase|Chondroitin polymerizing factor 2|Chondroitin sulfate glucuronyltransferase|Chondroitin synthase 3|ChPF-2|CHPF2|CHPF2_HUMAN|ChSy 3|ChSy-3|CSGLCA T|CSGlcA-T|N acetylgalactosaminyl proteoglycan 3 beta glucuronosyltransferase|N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: CHPF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human