Anti-RPA1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: RPA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Specifications
- Catalog No:
- 77797-766
- 77797-765
- Antigen Symbol:
- RPA1
- RPA1
- Antigen Name:
- Replication protein A 70 kDa DNA-binding subunit protein (2-616AA)
- Replication protein A 70 kDa DNA-binding subunit protein (2-616AA)
- Antigen Synonyms:
- Dmrpa1|Drosophila Replication Protein A|DRPA|HSSB|Human single stranded DNA binding protein|MST075|MSTP075|p70|REPA1|Replication factor A|Replication factor A protein 1|Replication protein A 70 kDa DNA-binding subunit|Replication protein A 70kDa DNA binding subunit|Replication protein A1 70kDa|Replication protein A1|RF A|RF-A protein 1|RFA|RFA1_HUMAN|RP A|RP-A p70|RPA 70|RPA|rpa1|Single stranded binding protein 70|Single-stranded DNA-binding protein
- Dmrpa1|Drosophila Replication Protein A|DRPA|HSSB|Human single stranded DNA binding protein|MST075|MSTP075|p70|REPA1|Replication factor A|Replication factor A protein 1|Replication protein A 70 kDa DNA-binding subunit|Replication protein A 70kDa DNA binding subunit|Replication protein A1 70kDa|Replication protein A1|RF A|RF-A protein 1|RFA|RFA1_HUMAN|RP A|RP-A p70|RPA 70|RPA|rpa1|Single stranded binding protein 70|Single-stranded DNA-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P27694
- P27694
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together