Anti-RASSF7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: RASSF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77818-670
- 77818-671
- Antigen Symbol:
- RASSF7
- RASSF7
- Antigen Name:
- Fusion protein of RASSF7
- Fusion protein of RASSF7
- Antigen Synonyms:
- C11orf13|Chromosome 11 open reading frame 13|HRAS1|HRAS1 related cluster 1|HRAS1 related cluster protein 1|HRAS1-related cluster protein 1|HRC1|MGC126069|MGC126070|Ras association (RalGDS/AF 6) domain family (N terminal) member 7|Ras association (RalGDS/AF 6) domain family 7|Ras association domain containing protein 7|Ras association domain-containing protein 7|RASF7_HUMAN|RASSF 7|RASSF7
- C11orf13|Chromosome 11 open reading frame 13|HRAS1|HRAS1 related cluster 1|HRAS1 related cluster protein 1|HRAS1-related cluster protein 1|HRC1|MGC126069|MGC126070|Ras association (RalGDS/AF 6) domain family (N terminal) member 7|Ras association (RalGDS/AF 6) domain family 7|Ras association domain containing protein 7|Ras association domain-containing protein 7|RASF7_HUMAN|RASSF 7|RASSF7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02833
- Q02833
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together