Anti-TNMD Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TNMD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-267
- 77803-267
- Antigen Symbol:
- TNMD
- TNMD
- Antigen Name:
- Tenomodulin protein (131-254AA)
- Tenomodulin protein (131-254AA)
- Antigen Synonyms:
- BRICD4|BRICHOS domain containing 4|CHM1 LIKE|ChM1L|Chondromodulin I like|Chondromodulin I like protein|Chondromodulin-1-like protein|Chondromodulin-I-like protein|hChM1L|hTeM|Myodulin|TeM|Tendin|Tenomodulin|Tenomodulin protein|Tnmd|TNMD_HUMAN|UNQ771/PRO1565
- BRICD4|BRICHOS domain containing 4|CHM1 LIKE|ChM1L|Chondromodulin I like|Chondromodulin I like protein|Chondromodulin-1-like protein|Chondromodulin-I-like protein|hChM1L|hTeM|Myodulin|TeM|Tendin|Tenomodulin|Tenomodulin protein|Tnmd|TNMD_HUMAN|UNQ771/PRO1565
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2S6
- Q9H2S6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together