Anti-FOXC2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FOXC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-507
- 77834-506
- Antigen Symbol:
- FOXC2
- FOXC2
- Antigen Name:
- Forkhead box protein C2 protein (24-294AA)
- Forkhead box protein C2 protein (24-294AA)
- Antigen Synonyms:
- Drosphilia Forkhead Homolog Like 14|FKHL 14|FKHL14|Forkhead Box C2|Forkhead box protein C2|Forkhead related protein FKHL14|Forkhead-related protein FKHL14|FOX C2|Foxc2|FOXC2_HUMAN|LD|Mesenchyme fork head protein 1|Mesenchyme Forkhead 1|MFH 1|MFH 1 protein|MFH-1 protein|MFH1|Transcription factor FKH 14|Transcription factor FKH-14
- Drosphilia Forkhead Homolog Like 14|FKHL 14|FKHL14|Forkhead Box C2|Forkhead box protein C2|Forkhead related protein FKHL14|Forkhead-related protein FKHL14|FOX C2|Foxc2|FOXC2_HUMAN|LD|Mesenchyme fork head protein 1|Mesenchyme Forkhead 1|MFH 1|MFH 1 protein|MFH-1 protein|MFH1|Transcription factor FKH 14|Transcription factor FKH-14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99958
- Q99958
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together