Anti-TM9SF3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TM9SF3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77802-776
- 77802-777
- Antigen Symbol:
- TM9SF3
- TM9SF3
- Antigen Name:
- Transmembrane 9 superfamily member 3 protein (133-300AA)
- Transmembrane 9 superfamily member 3 protein (133-300AA)
- Antigen Synonyms:
- Endomembrane protein emp70 precursor isolog|EP70 P iso|EP70-P-iso|RGD1564625|RP11 34E5.1|SM 11044 binding protein|SM-11044-binding protein|SMBP|TM9S3_HUMAN|Transmembrane 9 superfamily member 3|Transmembrane 9 superfamily protein member 3|UNQ245/PRO282
- Endomembrane protein emp70 precursor isolog|EP70 P iso|EP70-P-iso|RGD1564625|RP11 34E5.1|SM 11044 binding protein|SM-11044-binding protein|SMBP|TM9S3_HUMAN|Transmembrane 9 superfamily member 3|Transmembrane 9 superfamily protein member 3|UNQ245/PRO282
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HD45
- Q9HD45
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together