Anti-PI3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PI3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-084
- 77794-085
- Antigen Symbol:
- PI3
- PI3
- Antigen Name:
- Elafin protein (61-117AA)
- Elafin protein (61-117AA)
- Antigen Synonyms:
- Cementoin|ELAF_HUMAN|Elafin|Elafin/Skalp|Elastase Specific Inhibitor|Elastase-specific inhibitor|ES 1|ESI|Peptidase inhibitor 3|skin derived|PI 3|PI-3|PI3|Pre elafin|Protease inhibitor 3 skin derived |Protease inhibitor 3|skin derived (SKALP)|Protease inhibitor WAP3|SKALP|Skin derived Anti leukoproteinase|Skin-derived antileukoproteinase|Trappin 2|WAP four disulfide core domain 14|WAP four disulfide core domain protein 14|WAP four-disulfide core domain protein 14|WAP3|WFDC14
- Cementoin|ELAF_HUMAN|Elafin|Elafin/Skalp|Elastase Specific Inhibitor|Elastase-specific inhibitor|ES 1|ESI|Peptidase inhibitor 3|skin derived|PI 3|PI-3|PI3|Pre elafin|Protease inhibitor 3 skin derived |Protease inhibitor 3|skin derived (SKALP)|Protease inhibitor WAP3|SKALP|Skin derived Anti leukoproteinase|Skin-derived antileukoproteinase|Trappin 2|WAP four disulfide core domain 14|WAP four disulfide core domain protein 14|WAP four-disulfide core domain protein 14|WAP3|WFDC14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19957
- P19957
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together