Anti-RNASE2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RNASE2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-391
- 77797-390
- Antigen Symbol:
- RNASE2
- RNASE2
- Antigen Name:
- Non-secretory ribonuclease protein (28-161AA)
- Non-secretory ribonuclease protein (28-161AA)
- Antigen Synonyms:
- RNASE2|EDN|RNS2|Non-secretory ribonuclease|EC 4.6.1.18|Eosinophil-derived neurotoxin|RNase UpI-2|Ribonuclease 2|RNase 2|Ribonuclease US
- RNASE2|EDN|RNS2|Non-secretory ribonuclease|EC 4.6.1.18|Eosinophil-derived neurotoxin|RNase UpI-2|Ribonuclease 2|RNase 2|Ribonuclease US
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10153
- P10153
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together