Anti-gag Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: gag
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Moloney murine leukemia virus
Specifications
- Catalog No:
- 77816-875
- 77816-876
- Antigen Symbol:
- gag
- gag
- Antigen Name:
- Recombinant Moloney murine leukemia virus Gag polyprotein protein (216-478AA)
- Recombinant Moloney murine leukemia virus Gag polyprotein protein (216-478AA)
- Antigen Synonyms:
- gag| Gag polyprotein| Pr65gag| Core polyprotein) [Cleaved into: Matrix protein p15| MA)| RNA-binding phosphoprotein p12| pp12)| Capsid protein p30| CA)| Nucleocapsid protein p10-Gag| NC-gag)]
- gag| Gag polyprotein| Pr65gag| Core polyprotein) [Cleaved into: Matrix protein p15| MA)| RNA-binding phosphoprotein p12| pp12)| Capsid protein p30| CA)| Nucleocapsid protein p10-Gag| NC-gag)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Moloney murine leukemia virus
- Moloney murine leukemia virus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P03332
- P03332
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together