Anti-DVL3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DVL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-865
- 77833-864
- Antigen Symbol:
- DVL3
- DVL3
- Antigen Name:
- Segment polarity protein dishevelled homolog DVL-3 protein (509-696AA)
- Segment polarity protein dishevelled homolog DVL-3 protein (509-696AA)
- Antigen Synonyms:
- Dishevelled 3 (homologous to Drosophila dsh) |Dishevelled 3|dsh homolog (Drosophila)|Dishevelled|dsh homolog 3 (Drosophila)|Dishevelled-3|DSH homolog 3|dvl3|DVL3_HUMAN|KIAA0208|OTTHUMP00000210777|OTTHUMP00000210778|Segment polarity protein dishevelled homolog DVL 3|Segment polarity protein dishevelled homolog DVL-3
- Dishevelled 3 (homologous to Drosophila dsh) |Dishevelled 3|dsh homolog (Drosophila)|Dishevelled|dsh homolog 3 (Drosophila)|Dishevelled-3|DSH homolog 3|dvl3|DVL3_HUMAN|KIAA0208|OTTHUMP00000210777|OTTHUMP00000210778|Segment polarity protein dishevelled homolog DVL 3|Segment polarity protein dishevelled homolog DVL-3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92997
- Q92997
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together