Anti-IDH3B Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IDH3B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77808-723
- 77808-724
- Antigen Symbol:
- IDH3B
- IDH3B
- Antigen Name:
- Isocitrate dehydrogenase [NAD] subunit beta, mitochondrial protein (35-385AA)
- Isocitrate dehydrogenase [NAD] subunit beta, mitochondrial protein (35-385AA)
- Antigen Synonyms:
- FLJ11043|H-IDHB|Idh3B|IDH3B_HUMAN|Isocitrate dehydrogenase [NAD] subunit beta|mitochondrial |isocitrate dehydrogenase 3|beta subunit |Isocitric dehydrogenase|Isocitric dehydrogenase subunit beta|MGC903|mitochondrial|NAD(+)-specific ICDH|NAD(+)-specific ICDH subunit beta|NAD+-specific isocitrate dehydrogenase b subunit |NAD+-specific isocitrate dehydrogenase beta |OTTHUMP00000030023|OTTHUMP00000030024|RP46
- FLJ11043|H-IDHB|Idh3B|IDH3B_HUMAN|Isocitrate dehydrogenase [NAD] subunit beta|mitochondrial |isocitrate dehydrogenase 3|beta subunit |Isocitric dehydrogenase|Isocitric dehydrogenase subunit beta|MGC903|mitochondrial|NAD(+)-specific ICDH|NAD(+)-specific ICDH subunit beta|NAD+-specific isocitrate dehydrogenase b subunit |NAD+-specific isocitrate dehydrogenase beta |OTTHUMP00000030023|OTTHUMP00000030024|RP46
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43837
- O43837
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together