Anti-RYK Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: RYK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-409
- 77798-410
- Antigen Symbol:
- RYK
- RYK
- Antigen Name:
- Tyrosine-protein kinase RYK protein (494-586AA)
- Tyrosine-protein kinase RYK protein (494-586AA)
- Antigen Synonyms:
- D3S3195|ERK 3|Hydroxyaryl protein kinase|JTK 5|JTK 5A|JTK5|JTK5A|JTK5A protein tyrosine kinase|receptor like tyrosine kinase|receptor like tyrosine kinase precursor|RYK 1|Ryk|RYK_HUMAN|RYK1|Tyrosine protein kinase RYK|Tyrosine protein kinase RYK precursor|Tyrosine-protein kinase RYK|Vik
- D3S3195|ERK 3|Hydroxyaryl protein kinase|JTK 5|JTK 5A|JTK5|JTK5A|JTK5A protein tyrosine kinase|receptor like tyrosine kinase|receptor like tyrosine kinase precursor|RYK 1|Ryk|RYK_HUMAN|RYK1|Tyrosine protein kinase RYK|Tyrosine protein kinase RYK precursor|Tyrosine-protein kinase RYK|Vik
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P34925
- P34925
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together