Anti-UBR3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:500 to 1:1000, IF: 1:200 to 1:500
Type:
Antigen: UBR3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-997
- 77830-998
- Antigen Symbol:
- UBR3
- UBR3
- Antigen Name:
- E3 ubiquitin-protein ligase UBR3 protein (960-1113AA)
- E3 ubiquitin-protein ligase UBR3 protein (960-1113AA)
- Antigen Synonyms:
- DKFZp434P117|DKFZp686N10185|E3 ubiquitin-protein ligase UBR3|FLJ37422|KIAA2024|MGC126489|N-recognin-3|Ubiquitin-protein ligase E3-alpha-3|Ubiquitin-protein ligase E3-alpha-III|Ubr3|UBR3_HUMAN|Zinc finger protein 650|ZNF 650|ZNF650
- DKFZp434P117|DKFZp686N10185|E3 ubiquitin-protein ligase UBR3|FLJ37422|KIAA2024|MGC126489|N-recognin-3|Ubiquitin-protein ligase E3-alpha-3|Ubiquitin-protein ligase E3-alpha-III|Ubr3|UBR3_HUMAN|Zinc finger protein 650|ZNF 650|ZNF650
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZT12
- Q6ZT12
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together