Anti-FKBP14 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FKBP14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-578
- 77836-577
- Antigen Symbol:
- FKBP14
- FKBP14
- Antigen Name:
- Peptidyl-prolyl cis-trans isomerase FKBP14 protein (20-211AA)
- Peptidyl-prolyl cis-trans isomerase FKBP14 protein (20-211AA)
- Antigen Synonyms:
- 22 kDa FK506 binding protein| 22 kDa FK506-binding protein| 22 kDa FKBP| FK506 binding protein 14 (22 kDa)| FK506 binding protein 14| FK506-binding protein 14| FKB14_HUMAN| FKBP 22| FKBP-14| FKBP-22| FKBP14| FKBP22| FLJ20731| Peptidyl prolyl cis trans isomerase| Peptidyl-prolyl cis-trans isomerase FKBP14| PPIase| PPIase FKBP14| Rotamase
- 22 kDa FK506 binding protein| 22 kDa FK506-binding protein| 22 kDa FKBP| FK506 binding protein 14 (22 kDa)| FK506 binding protein 14| FK506-binding protein 14| FKB14_HUMAN| FKBP 22| FKBP-14| FKBP-22| FKBP14| FKBP22| FLJ20731| Peptidyl prolyl cis trans isomerase| Peptidyl-prolyl cis-trans isomerase FKBP14| PPIase| PPIase FKBP14| Rotamase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NWM8
- Q9NWM8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together