Anti-KDM6A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: KDM6A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-189
- 77804-190
- Antigen Symbol:
- KDM6A
- KDM6A
- Antigen Name:
- Lysine-specific demethylase 6A protein (620-749AA)
- Lysine-specific demethylase 6A protein (620-749AA)
- Antigen Synonyms:
- bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)|bA386N14.2|bA386N14.2 ubiquitously transcribed X chromosome tetratricopeptide repeat protein UTX|DKFZp686A03225|Histone demethylase UTX|KABUK2|KDM6A_HUMAN|Lysine (K) specific demethylase 6A|Lysine K specific demethylase 6A|Lysine specific demethylase 6A ubiquitously transcribed tetratricopeptide repeat gene X chromosome|Lysine-specific demethylase 6A|MGC141941|Ubiquitously transcribed tetratricopeptide repeat X chromosome|Ubiquitously transcribed tetratricopeptide repeat protein X linked|Ubiquitously transcribed TPR gene on the X chromosome|Ubiquitously transcribed TPR protein on the X chromosome|Ubiquitously transcribed X chromosome tetratricopeptide repeat protein|Ubiquitously-transcribed TPR protein on the X chromosome|Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein|UTX
- bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)|bA386N14.2|bA386N14.2 ubiquitously transcribed X chromosome tetratricopeptide repeat protein UTX|DKFZp686A03225|Histone demethylase UTX|KABUK2|KDM6A_HUMAN|Lysine (K) specific demethylase 6A|Lysine K specific demethylase 6A|Lysine specific demethylase 6A ubiquitously transcribed tetratricopeptide repeat gene X chromosome|Lysine-specific demethylase 6A|MGC141941|Ubiquitously transcribed tetratricopeptide repeat X chromosome|Ubiquitously transcribed tetratricopeptide repeat protein X linked|Ubiquitously transcribed TPR gene on the X chromosome|Ubiquitously transcribed TPR protein on the X chromosome|Ubiquitously transcribed X chromosome tetratricopeptide repeat protein|Ubiquitously-transcribed TPR protein on the X chromosome|Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein|UTX
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15550
- O15550
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together