Anti-DCSTAMP Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DCSTAMP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77802-767
- 77802-766
- Antigen Symbol:
- DCSTAMP
- DCSTAMP
- Antigen Name:
- Dendritic cell-specific transmembrane protein (119-209AA)
- Dendritic cell-specific transmembrane protein (119-209AA)
- Antigen Synonyms:
- DC-STAMP|DCSTP_HUMAN|Dendritic cell-specific transmembrane protein|Dendrocyte-expressed seven transmembrane protein|FIND|hDC-STAMP|IL-Four INDuced|IL-four-induced protein|Transmembrane 7 superfamily member 4
- DC-STAMP|DCSTP_HUMAN|Dendritic cell-specific transmembrane protein|Dendrocyte-expressed seven transmembrane protein|FIND|hDC-STAMP|IL-Four INDuced|IL-four-induced protein|Transmembrane 7 superfamily member 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H295
- Q9H295
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together