Anti-TNFAIP8 Rabbit Polyclonal Antibody
Catalog # 77811-215
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Tumor necrosis factor alpha-induced protein 8
- Antigen Symbol:TNFAIP8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O95379
- Antigen Synonyms:GG2 1|Head and neck tumor and metastasis-related protein|MDC-3.13|NDED|NF-kappa-B-inducible DED-containing protein|SCC S2|SCC-S2|SCCS2|TFIP8_HUMAN|TNF alpha-induced protein 8|TNF induced protein|TNF-induced protein GG2-1|TNFAIP 8|TNFAIP8|Tumor necrosis factor alpha induced protein 8|Tumor necrosis factor alpha-induced protein 8
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TNFAIP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat