Anti-PSMD4 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PSMD4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-881
- 77795-880
- Antigen Symbol:
- PSMD4
- PSMD4
- Antigen Name:
- 26S proteasome non-ATPase regulatory subunit 4 protein (161-377AA)
- 26S proteasome non-ATPase regulatory subunit 4 protein (161-377AA)
- Antigen Synonyms:
- 26S protease subunit S5a|26S proteasome non ATPase regulatory subunit 4|26S proteasome non-ATPase regulatory subunit 4|26S proteasome regulatory subunit rpn10|26S proteasome regulatory subunit S5A|AF 1|AF|AF1|Angiocidin|Antisecretory factor 1|ASF|DS5a|MCB 1|MCB1|Multiubiquitin chain binding protein|Multiubiquitin chain-binding protein|OTTHUMP00000059963|Prosome macropain|Proteasome (prosome macropain) 26S subunit non ATPase 4|Proteasome 19S S5A|Proteasome 26S non ATPase subunit 4|Proteasome 26S subunit non ATPase 4|PSMD 4|Psmd4|PSMD4_HUMAN|pUB R5|pUBR5|Rpn 10|Rpn10|RPN10 homolog|S5A|S5a/antisecretory factor protein
- 26S protease subunit S5a|26S proteasome non ATPase regulatory subunit 4|26S proteasome non-ATPase regulatory subunit 4|26S proteasome regulatory subunit rpn10|26S proteasome regulatory subunit S5A|AF 1|AF|AF1|Angiocidin|Antisecretory factor 1|ASF|DS5a|MCB 1|MCB1|Multiubiquitin chain binding protein|Multiubiquitin chain-binding protein|OTTHUMP00000059963|Prosome macropain|Proteasome (prosome macropain) 26S subunit non ATPase 4|Proteasome 19S S5A|Proteasome 26S non ATPase subunit 4|Proteasome 26S subunit non ATPase 4|PSMD 4|Psmd4|PSMD4_HUMAN|pUB R5|pUBR5|Rpn 10|Rpn10|RPN10 homolog|S5A|S5a/antisecretory factor protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55036
- P55036
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together