Anti-ADGRL4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ADGRL4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-559
- 77838-560
- Antigen Symbol:
- ADGRL4
- ADGRL4
- Antigen Name:
- Adhesion G protein-coupled receptor L4 protein (85-299AA)
- Adhesion G protein-coupled receptor L4 protein (85-299AA)
- Antigen Synonyms:
- ADGRL4|Adhesion G protein-coupled receptor L4|EGF|EGF latrophilin and seven transmembrane domain containing 1|EGF TM7 latrophilin related protein|latrophilin and seven transmembrane domain-containing protein 1|latrophilin|and 7 transmembrane domain-containing protein 1|EGF-|TM7-|and latrophilin-related protein|EGF-TM7-latrophilin-related protein|EGFTM7 latrophilin related protein|ELTD 1|ELTD1|ELTD1_HUMAN|ETL|ETL protein|KPG_003|UNQ202/PRO228|zgc:63629
- ADGRL4|Adhesion G protein-coupled receptor L4|EGF|EGF latrophilin and seven transmembrane domain containing 1|EGF TM7 latrophilin related protein|latrophilin and seven transmembrane domain-containing protein 1|latrophilin|and 7 transmembrane domain-containing protein 1|EGF-|TM7-|and latrophilin-related protein|EGF-TM7-latrophilin-related protein|EGFTM7 latrophilin related protein|ELTD 1|ELTD1|ELTD1_HUMAN|ETL|ETL protein|KPG_003|UNQ202/PRO228|zgc:63629
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HBW9
- Q9HBW9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together