Anti-SULT1A2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SULT1A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-548
- 77801-547
- Antigen Symbol:
- SULT1A2
- SULT1A2
- Antigen Name:
- Sulfotransferase 1A2 protein (195-284AA)
- Sulfotransferase 1A2 protein (195-284AA)
- Antigen Synonyms:
- Aryl sulfotransferase 2|Arylamine sulfotransferase|HAST4|MGC142287|MGC142289|P PST|P-PST 2|Phenol preferring phenol sulfotransferase 2|Phenol sulfotransferase 2|Phenol-sulfating phenol sulfotransferase 2|Phenolic metabolizing (P) form of PST|ST1A2|ST1A2_HUMAN|STP2|Sulfotransferase 1A2|Sulfotransferase family cytosolic 1A phenol preferring member 2|SULT1A2|Thermostable phenol sulfotransferase|TSPST2
- Aryl sulfotransferase 2|Arylamine sulfotransferase|HAST4|MGC142287|MGC142289|P PST|P-PST 2|Phenol preferring phenol sulfotransferase 2|Phenol sulfotransferase 2|Phenol-sulfating phenol sulfotransferase 2|Phenolic metabolizing (P) form of PST|ST1A2|ST1A2_HUMAN|STP2|Sulfotransferase 1A2|Sulfotransferase family cytosolic 1A phenol preferring member 2|SULT1A2|Thermostable phenol sulfotransferase|TSPST2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P50226
- P50226
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together