Anti-TFF3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TFF3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77802-336
- 77802-335
- Antigen Symbol:
- TFF3
- TFF3
- Antigen Name:
- Trefoil factor 3 protein (22-73AA)
- Trefoil factor 3 protein (22-73AA)
- Antigen Synonyms:
- hITF|hP1.B|Intestinal trefoil factor|ITF|mITF|OTTMUSP00000021729|P1B|Polypeptide P1.B|TFF3|TFF3_HUMAN|TFI|TREFOIL|Trefoil factor (intestinal)|Trefoil factor 3
- hITF|hP1.B|Intestinal trefoil factor|ITF|mITF|OTTMUSP00000021729|P1B|Polypeptide P1.B|TFF3|TFF3_HUMAN|TFI|TREFOIL|Trefoil factor (intestinal)|Trefoil factor 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q07654
- Q07654
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together