Anti-PSMA1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: PSMA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77795-745
- 77795-746
- Antigen Symbol:
- PSMA1
- PSMA1
- Antigen Name:
- Proteasome subunit alpha type-1 protein (163-263AA)
- Proteasome subunit alpha type-1 protein (163-263AA)
- Antigen Synonyms:
- 30 kDa prosomal protein| HC 2| HC2| Macropain subunit C2| Macropain subunit nu| MGC14542| MGC14575| MGC14751| MGC1667| MGC21459| MGC22853| MGC23915| Multicatalytic endopeptidase complex subunit C2| NU| PROS 30| PROS-30| PROS30| Proteasome (prosome macropain) subunit alpha type 1| Proteasome alpha 1 subunit| Proteasome component C2| Proteasome nu chain| Proteasome subunit alpha type 1| Proteasome subunit alpha type I| Proteasome subunit alpha type-1| Proteasome subunit nu| Protein P30 33K| PSA1_HUMAN| PSC 2| PSC2| PSMA 1| psmA1
- 30 kDa prosomal protein| HC 2| HC2| Macropain subunit C2| Macropain subunit nu| MGC14542| MGC14575| MGC14751| MGC1667| MGC21459| MGC22853| MGC23915| Multicatalytic endopeptidase complex subunit C2| NU| PROS 30| PROS-30| PROS30| Proteasome (prosome macropain) subunit alpha type 1| Proteasome alpha 1 subunit| Proteasome component C2| Proteasome nu chain| Proteasome subunit alpha type 1| Proteasome subunit alpha type I| Proteasome subunit alpha type-1| Proteasome subunit nu| Protein P30 33K| PSA1_HUMAN| PSC 2| PSC2| PSMA 1| psmA1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25786
- P25786
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together