Anti-STK26 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: STK26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77812-855
- 77812-856
- Antigen Symbol:
- STK26
- STK26
- Antigen Name:
- Synthetic peptide of STK26
- Synthetic peptide of STK26
- Antigen Synonyms:
- 2610018G03Rik|EC 2.7.11.1|Mammalian Ste20 like protein kinase 4|Mammalian STE20-like protein kinase 4|Mammalian sterile 20 like 4|MASK|MST 4|MST-4|MST3 and SOK1 related kinase|Mst3 and SOK1-related kinase|Mst4|MST4_HUMAN|RGD1563568|RP23-245F8.1|RP6 213H19.1|Serine/threonine protein kinase 26|Serine/threonine protein kinase MASK|Serine/threonine protein kinase MST 4|Serine/threonine protein kinase MST4|Serine/threonine-protein kinase MASK|Serine/threonine-protein kinase MST4|STE20 like kinase MST 4|STE20 like kinase MST4|STE20-like kinase MST4|STK26
- 2610018G03Rik|EC 2.7.11.1|Mammalian Ste20 like protein kinase 4|Mammalian STE20-like protein kinase 4|Mammalian sterile 20 like 4|MASK|MST 4|MST-4|MST3 and SOK1 related kinase|Mst3 and SOK1-related kinase|Mst4|MST4_HUMAN|RGD1563568|RP23-245F8.1|RP6 213H19.1|Serine/threonine protein kinase 26|Serine/threonine protein kinase MASK|Serine/threonine protein kinase MST 4|Serine/threonine protein kinase MST4|Serine/threonine-protein kinase MASK|Serine/threonine-protein kinase MST4|STE20 like kinase MST 4|STE20 like kinase MST4|STE20-like kinase MST4|STK26
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P289
- Q9P289
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together