Anti-SDHAF1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SDHAF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77818-124
- 77818-125
- Antigen Symbol:
- SDHAF1
- SDHAF1
- Antigen Name:
- Succinate dehydrogenase assembly factor 1, mitochondrial protein (78-115AA)
- Succinate dehydrogenase assembly factor 1, mitochondrial protein (78-115AA)
- Antigen Synonyms:
- LYR motif containing 8|LYR motif-containing protein 8|LYRM8|mitochondrial|SDH assembly factor 1|SDHAF1|SDHF1_HUMAN|Succinate dehydrogenase assembly factor 1|Succinate dehydrogenase assembly factor 1 mitochondrial|Succinate dehydrogenase complex assembly factor 1
- LYR motif containing 8|LYR motif-containing protein 8|LYRM8|mitochondrial|SDH assembly factor 1|SDHAF1|SDHF1_HUMAN|Succinate dehydrogenase assembly factor 1|Succinate dehydrogenase assembly factor 1 mitochondrial|Succinate dehydrogenase complex assembly factor 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A6NFY7
- A6NFY7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together