Anti-DIXDC1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DIXDC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-507
- 77824-506
- Antigen Symbol:
- DIXDC1
- DIXDC1
- Antigen Name:
- Dixin protein (70-333AA)
- Dixin protein (70-333AA)
- Antigen Synonyms:
- CCD1| Ccd1Aalpha1L| Ccd1Abeta1L| Ccd1BalphaL| Ccd1BbetaL| Ccd1CL| Coiled coil DIX1| Coiled coil protein DIX1| Coiled-coil protein DIX1| Coiled-coil-DIX1| DIX domain containing 1| DIX domain containing protein 1| DIX domain-containing protein 1| DIXC1_HUMAN| DIXDC1| Dixin| KIAA1735
- CCD1| Ccd1Aalpha1L| Ccd1Abeta1L| Ccd1BalphaL| Ccd1BbetaL| Ccd1CL| Coiled coil DIX1| Coiled coil protein DIX1| Coiled-coil protein DIX1| Coiled-coil-DIX1| DIX domain containing 1| DIX domain containing protein 1| DIX domain-containing protein 1| DIXC1_HUMAN| DIXDC1| Dixin| KIAA1735
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q155Q3
- Q155Q3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together