Anti-UBTD2 Rabbit Polyclonal Antibody
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:30 to 1:150
Type:
Antigen: UBTD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:30 to 1:150
Type:
Antigen: UBTD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77818-025
- 77818-026
- Antigen Symbol:
- UBTD2
- UBTD2
- Antigen Name:
- Synthetic peptide of UBTD2
- Synthetic peptide of UBTD2
- Antigen Synonyms:
- ubiquitin domain containing 2
- ubiquitin domain containing 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human|Mouse
- Human|Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WUN7
- Q8WUN7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Product Family Options
- SizePackaging
- : A51892-100UL
Specifications
Cat. No.77818-025Antigen SymbolUBTD2Antigen NameSynthetic peptide of UBTD2Antigen Synonymsubiquitin domain containing 2ClonalityPolyclonalConjugationUnconjugatedReactivityHuman|MouseHostRabbitGene IDQ8WUN7IsotypeIgGELISAYesImmunoChemistryYesFormLiquidPurificationAntigen affinity purifiedStorage BufferPBS, 0.05% sodium azide, 40% glycerol, pH 7.4Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice - : A51892-50UL
Specifications
Cat. No.77818-026Antigen SymbolUBTD2Antigen NameSynthetic peptide of UBTD2Antigen Synonymsubiquitin domain containing 2ClonalityPolyclonalConjugationUnconjugatedReactivityHuman|MouseHostRabbitGene IDQ8WUN7IsotypeIgGELISAYesImmunoChemistryYesFormLiquidPurificationAntigen affinity purifiedStorage BufferPBS, 0.05% sodium azide, 40% glycerol, pH 7.4Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice



