Anti-MECOM Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
About this item
Recommended dilution: IHC: 1:500 to 1:1000
Type:
Antigen: MECOM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recommended dilution: IHC: 1:500 to 1:1000
Type:
Antigen: MECOM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications for Product Family
Specifications
- Catalog No:
- 77824-319
- 77824-320
- Antigen Symbol:
- MECOM
- MECOM
- Antigen Name:
- Histone-lysine N-methyltransferase MECOM protein (534-632AA)
- Histone-lysine N-methyltransferase MECOM protein (534-632AA)
- Antigen Synonyms:
- MECOM|EVI1|MDS1|PRDM3|Histone-lysine N-methyltransferase MECOM|EC 2.1.1.-|Ecotropic virus integration site 1 protein homolog|EVI-1|MDS1 and EVI1 complex locus protein|Myelodysplasia syndrome 1 protein|Myelodysplasia syndrome-associated protein 1
- MECOM|EVI1|MDS1|PRDM3|Histone-lysine N-methyltransferase MECOM|EC 2.1.1.-|Ecotropic virus integration site 1 protein homolog|EVI-1|MDS1 and EVI1 complex locus protein|Myelodysplasia syndrome 1 protein|Myelodysplasia syndrome-associated protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q03112
- Q03112
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Product Family Options
Product Information
- SizePackagingAvailabilityPrice
- Supplier Number: A55408-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77824-319Antigen SymbolMECOMAntigen NameHistone-lysine N-methyltransferase MECOM protein (534-632AA)Antigen SynonymsMECOM|EVI1|MDS1|PRDM3|Histone-lysine N-methyltransferase MECOM|EC 2.1.1.-|Ecotropic virus integration site 1 protein homolog|EVI-1|MDS1 and EVI1 complex locus protein|Myelodysplasia syndrome 1 protein|Myelodysplasia syndrome-associated protein 1ClonalityRecombinant PolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene IDQ03112IsotypeIgGELISAYesImmunoChemistryYesFormLiquidPurification>95%, Protein G purifiedStorage Buffer0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice - Supplier Number: A55408-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77824-320Antigen SymbolMECOMAntigen NameHistone-lysine N-methyltransferase MECOM protein (534-632AA)Antigen SynonymsMECOM|EVI1|MDS1|PRDM3|Histone-lysine N-methyltransferase MECOM|EC 2.1.1.-|Ecotropic virus integration site 1 protein homolog|EVI-1|MDS1 and EVI1 complex locus protein|Myelodysplasia syndrome 1 protein|Myelodysplasia syndrome-associated protein 1ClonalityRecombinant PolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene IDQ03112IsotypeIgGELISAYesImmunoChemistryYesFormLiquidPurification>95%, Protein G purifiedStorage Buffer0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



