Anti-COX16 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:500 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: COX16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77826-680
- 77826-681
- Antigen Symbol:
- COX16
- COX16
- Antigen Name:
- Fusion protein of COX16
- Fusion protein of COX16
- Antigen Synonyms:
- C14orf112||cytochrome c oxidase assembly homolog (S. cerevisiae)|_HUMAN|Cytochrome c oxidase assembly factor|Cytochrome c oxidase assembly protein homolog|HSPC203|mitochondrial|PTD019
- C14orf112||cytochrome c oxidase assembly homolog (S. cerevisiae)|_HUMAN|Cytochrome c oxidase assembly factor|Cytochrome c oxidase assembly protein homolog|HSPC203|mitochondrial|PTD019
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P0S2
- Q9P0S2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together