Anti-THOC7 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: THOC7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-981
- 77829-980
- Antigen Symbol:
- THOC7
- THOC7
- Antigen Name:
- THO complex subunit 7 homolog protein (4-204AA)
- THO complex subunit 7 homolog protein (4-204AA)
- Antigen Synonyms:
- FLJ23445|fSAP24|Functional spliceosome associated protein 24|Functional spliceosome-associated protein 24|hTREX30|Ngg1 interacting factor 3 like 1 binding protein 1|Ngg1 interacting factor 3 like protein 1 binding protein 1|Ngg1-interacting factor 3-like protein 1-binding protein 1|NIF3L1 binding protein 1|NIF3L1-binding protein 1|NIF3L1BP1|THO complex 7 homolog (Drosophila)|THO complex 7 homolog|THO complex subunit 7 homolog|THOC 7|thoc7|THOC7_HUMAN
- FLJ23445|fSAP24|Functional spliceosome associated protein 24|Functional spliceosome-associated protein 24|hTREX30|Ngg1 interacting factor 3 like 1 binding protein 1|Ngg1 interacting factor 3 like protein 1 binding protein 1|Ngg1-interacting factor 3-like protein 1-binding protein 1|NIF3L1 binding protein 1|NIF3L1-binding protein 1|NIF3L1BP1|THO complex 7 homolog (Drosophila)|THO complex 7 homolog|THO complex subunit 7 homolog|THOC 7|thoc7|THOC7_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6I9Y2
- Q6I9Y2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together