Anti-RPL14 Rabbit Polyclonal Antibody
Catalog # 77810-461
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the Internal region of Ribosomal Protein L14
- Antigen Symbol:RPL14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P50914
- Antigen Synonyms:3100001N19Rik|60S ribosomal protein L14|AA407502|AL022816|CAG ISL 7|CAG-ISL 7|CTG B33|hRL 14|hRL14|L14|MGC88594|Ribosomal protein L14|RL 14|RL14|RL14_HUMAN|RP L14|RPL 14|rpl14
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:5000
Type:
Antigen: RPL14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat