Anti-CDK12 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: CDK12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-141
- 77841-142
- Antigen Symbol:
- CDK12
- CDK12
- Antigen Name:
- Cyclin-dependent kinase 12 protein (1309-1490AA)
- Cyclin-dependent kinase 12 protein (1309-1490AA)
- Antigen Synonyms:
- arginine/serine-rich| Cdc2 related kinase arginine/serine rich| CDC2 related protein kinase 7| Cdc2-related kinase| CDC2-related protein kinase 7| Cdk12| CDK12_HUMAN| Cell division cycle 2 related protein kinase 7| Cell division cycle 2-related protein kinase 7| Cell division protein kinase 12| CRK7| CRKR| CrkRS| Cyclin-dependent kinase 12| EC 2.7.11.22| hCDK12| kiaa0904| Pksc
- arginine/serine-rich| Cdc2 related kinase arginine/serine rich| CDC2 related protein kinase 7| Cdc2-related kinase| CDC2-related protein kinase 7| Cdk12| CDK12_HUMAN| Cell division cycle 2 related protein kinase 7| Cell division cycle 2-related protein kinase 7| Cell division protein kinase 12| CRK7| CRKR| CrkRS| Cyclin-dependent kinase 12| EC 2.7.11.22| hCDK12| kiaa0904| Pksc
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NYV4
- Q9NYV4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together