Anti-KLF5 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KLF5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-908
- 77821-907
- Antigen Symbol:
- KLF5
- KLF5
- Antigen Name:
- Krueppel-like factor 5 protein (69-346AA)
- Krueppel-like factor 5 protein (69-346AA)
- Antigen Synonyms:
- Basic transcription element binding protein 2|Basic transcription element-binding protein 2|BTE binding protein 2|BTE-binding protein 2|BTEB 2|BTEB2|CKLF|Colon krueppel like factor|Colon krueppel-like factor|GC box binding protein 2|GC-box-binding protein 2|IKLF|Intestinal enriched krueppel like factor|Intestinal-enriched krueppel-like factor|KLF 5|Klf5|KLF5_HUMAN|Krueppel like factor 5|Krueppel-like factor 5|Kruppel like factor 5 intestinal|Transcription factor BTEB 2|Transcription factor BTEB2
- Basic transcription element binding protein 2|Basic transcription element-binding protein 2|BTE binding protein 2|BTE-binding protein 2|BTEB 2|BTEB2|CKLF|Colon krueppel like factor|Colon krueppel-like factor|GC box binding protein 2|GC-box-binding protein 2|IKLF|Intestinal enriched krueppel like factor|Intestinal-enriched krueppel-like factor|KLF 5|Klf5|KLF5_HUMAN|Krueppel like factor 5|Krueppel-like factor 5|Kruppel like factor 5 intestinal|Transcription factor BTEB 2|Transcription factor BTEB2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13887
- Q13887
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together