Anti-CALML5 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: CALML5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77838-960
- 77838-959
- Antigen Symbol:
- CALML5
- CALML5
- Antigen Name:
- Calmodulin-like protein 5 protein (1-146AA)
- Calmodulin-like protein 5 protein (1-146AA)
- Antigen Synonyms:
- CALL5_HUMAN|CALML5|Calmodulin like 5|Calmodulin-like protein 5|Calmodulin-like skin protein|CLSP
- CALL5_HUMAN|CALML5|Calmodulin like 5|Calmodulin-like protein 5|Calmodulin-like skin protein|CLSP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZT1
- Q9NZT1
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together