Anti-RUNX1T1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: RUNX1T1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77828-645
- 77828-644
- Antigen Symbol:
- RUNX1T1
- RUNX1T1
- Antigen Name:
- Protein CBFA2T1 protein (416-525AA)
- Protein CBFA2T1 protein (416-525AA)
- Antigen Synonyms:
- Acute myelogenous leukemia 1 translocation 1 cyclin D related|Acute myelogenous leukemia 1 translocation 1 protein|AML1 MTG8|AML1T1|CBFA2T1|CDR|Core binding factor runt domain alpha subunit 2 translocated to 1|Core binding factor runt domain alpha subunit 2 translocated to 1 cyclin D related|Cyclin D related|Cyclin D related protein|Cyclin-D-related protein|Eight twenty one protein|ETO|ETO protein|MGC2796|MTG 8|MTG 8b|MTG8|MTG8 protein|MTG8_HUMAN|MTG8b|Myeloid translocation gene on 8q22|Protein CBFA2T1|Protein ETO|Protein MTG 8|Protein MTG8|Runt related transcription factor 1 translocated to 1 (cyclin D related)|Runt related transcription factor 1 translocated to 1|Runt related transcription factor 1 translocated to 1 cyclin D related|RUNX1 translocation partner 1|RUNX1T1|Zinc finger MYND domain containing protein 2|Zinc finger MYND domain-containing protein 2|ZMYND 2|ZMYND2
- Acute myelogenous leukemia 1 translocation 1 cyclin D related|Acute myelogenous leukemia 1 translocation 1 protein|AML1 MTG8|AML1T1|CBFA2T1|CDR|Core binding factor runt domain alpha subunit 2 translocated to 1|Core binding factor runt domain alpha subunit 2 translocated to 1 cyclin D related|Cyclin D related|Cyclin D related protein|Cyclin-D-related protein|Eight twenty one protein|ETO|ETO protein|MGC2796|MTG 8|MTG 8b|MTG8|MTG8 protein|MTG8_HUMAN|MTG8b|Myeloid translocation gene on 8q22|Protein CBFA2T1|Protein ETO|Protein MTG 8|Protein MTG8|Runt related transcription factor 1 translocated to 1 (cyclin D related)|Runt related transcription factor 1 translocated to 1|Runt related transcription factor 1 translocated to 1 cyclin D related|RUNX1 translocation partner 1|RUNX1T1|Zinc finger MYND domain containing protein 2|Zinc finger MYND domain-containing protein 2|ZMYND 2|ZMYND2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q06455
- Q06455
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together