Anti-DAZ2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DAZ2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-182
- 77825-181
- Antigen Symbol:
- DAZ2
- DAZ2
- Antigen Name:
- Deleted in azoospermia protein 2 protein (1-200AA)
- Deleted in azoospermia protein 2 protein (1-200AA)
- Antigen Synonyms:
- DAZ2|DAZ2_HUMAN|DAZ3|Deleted in azoospermia 2|Deleted in azoospermia protein 2|Deleted in azoospermia protein 3|MGC126442|pDP1678
- DAZ2|DAZ2_HUMAN|DAZ3|Deleted in azoospermia 2|Deleted in azoospermia protein 2|Deleted in azoospermia protein 3|MGC126442|pDP1678
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13117
- Q13117
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together