Anti-VSIR Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VSIR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-080
- 77842-081
- Antigen Symbol:
- VSIR
- VSIR
- Antigen Name:
- V-type immunoglobulin domain-containing suppressor of T-cell activation protein (37-194AA)
- V-type immunoglobulin domain-containing suppressor of T-cell activation protein (37-194AA)
- Antigen Synonyms:
- B7 H5|B7H5|C10orf54|chromosome 10 open reading frame 54|DD1alpha|GI24|GI24_HUMAN|PD-1H|PDCD1 homolog|Platelet receptor Gi24|PP2135|sisp 1|SISP1|stress induced secreted protein 1|UNQ730/PRO1412|V domain Ig suppressor of T cell activation|V set domain containing immunoregulatory receptor|V set immunoregulatory receptor|V type immunoglobulin domain containing suppressor of T cell activation|VISTA
- B7 H5|B7H5|C10orf54|chromosome 10 open reading frame 54|DD1alpha|GI24|GI24_HUMAN|PD-1H|PDCD1 homolog|Platelet receptor Gi24|PP2135|sisp 1|SISP1|stress induced secreted protein 1|UNQ730/PRO1412|V domain Ig suppressor of T cell activation|V set domain containing immunoregulatory receptor|V set immunoregulatory receptor|V type immunoglobulin domain containing suppressor of T cell activation|VISTA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H7M9
- Q9H7M9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together